missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Glucose, 98%

Product Code. 10025953
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 921-60-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00148913
InChI Key GZCGUPFRVQAUEE-VANKVMQKSA-N
Synonym l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer
PubChem CID 10954115
ChEBI CHEBI:37626
IUPAC Name (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 153.0°C to 156.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition − 52.00 (20.00°C c=10,H2O,NH3)
Specific Rotation − 52.00
Solubility Information (10% in water) Clear colorless
Formula Weight 180.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Glucose
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'