missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Glutamic acid, 99%

Product Code. 10502751 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic Drum
Unit Size:
100g
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 10502751

Brand: Thermo Scientific Chemicals 156211000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
Synonym l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
PubChem CID 33032
ChEBI CHEBI:16015
IUPAC Name (2S)-2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

Ammonium 200ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99%
Loss on Drying 0.5% (105°C, 3 hrs) max.
Other Amino Acids Not detectable
Packaging Plastic Bottle
Linear Formula HO2CCH2CH2CH(NH2)CO2H
Specific Rotation +30.5° to +32.5° (20°C, 589nm) (on dried substance) (c=10, 2N HCl)
Sulfate 300ppm max.
Sulfated Ash 0.1% max.
Sublimation Point >200
Specific Rotation Condition +32.00 (20.00°C c=10,2N HCl)
Quantity 100 g
Formula Weight 147.13
Percent Purity 98.5 to 100.5% (on dried substance)
Physical Form Powder
Chemical Name or Material L(+)-Glutamic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.