missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic Acid Ammonium Salt, MP Biomedicals™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
100 g
Specifications
CAS | 7558-63-6 |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Molecular Formula | C5H12N2O4 |
Synonym | unii-245k560gaw, ammonium glutamate, l-glutamic acid, monoammonium salt, ammonium l-glutamate, monoammonium l-glutamate, ammoniumglutaminat german, glutamic acid, monoammonium salt, l, l-glutamic acid amine, 56-86-0 parent, ammoniumglutaminat |
InChI Key | PHKGGXPMPXXISP-DFWYDOINSA-N |
SMILES | C(CC(=O)[O-])C(C(=O)O)N.[NH4+] |
IUPAC Name | azanium;(4S)-4-amino-5-hydroxy-5-oxopentanoate |
Molecular Weight (g/mol) | 164.161 |
PubChem CID | 89887156 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'