missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid di-tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.
Solubility
Soluble in methanol (50gmg/ml).
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.
Solubility
Soluble in methanol (50gmg/ml).
Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Beilstein | 3573738 |
Solubility Information | Soluble in methanol (50gmg/ml). |
Formula Weight | 295.8 |
Percent Purity | 98% |
Quantity | 1 g |
Physical Form | Solid |
Chemical Name or Material | L-Glutamic acid di-tert-butyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'