missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic Acid, Extra Pure, SLR, Fisher Chemical
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Specifications
Specifications
CAS | 56-86-0 |
Melting Point | 205°C |
Color | White |
pH | 3.3 |
Packaging | Plastic powder jar |
Molecular Formula | C5H9NO4 |
MDL Number | MFCD00002634 |
Quantity | 100 g |
Synonym | l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex |
InChI Key | WHUUTDBJXJRKMK-UHFFFAOYNA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'