missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid, Tocris Bioscience™
Click to view available options
Quantity:
1 g
Description
L-Glutamic acid is a the predominant excitatory transmitter in the mammalian central nervous system. L-Glutamic acid acts at ionotropic and metabotropic glutamate receptors.
MNI-caged-L-glutamate (Cat. No. 1490) also available.
Specifications
Specifications
CAS | 56-86-0 |
Molecular Formula | C5H9NO4 |
MDL Number | MFCD00002634 |
Synonym | l-glutamic acid, glutamic acid, 2s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex |
InChI Key | WHUUTDBJXJRKMK-UHFFFAOYNA-N |
SMILES | NC(CCC(O)=O)C(O)=O |
IUPAC Name | 2-aminopentanedioic acid |
Molecular Weight (g/mol) | 147.13 |
Quantity | 1 g |
PubChem CID | 33032 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction