missing translation for 'onlineSavingsMsg'
Learn More

Cytiva L-Glutamine

Supplement serum- and protein-free media using this amino acid.

Brand:  Cytiva SH30034.01

16.34 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 10453332

  • £25.90 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Format: Liquid
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store frozen
29.2 mg/mL
0.1 μm Sterile-filtered
L-Glutamine, 200mM Solution (29.2mg/mL-Glutamine in 0.85% NaCl), 100mL
Liquid
100 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers