missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva L-Glutamine
Supplement serum- and protein-free media using this amino acid.
Brand: Cytiva SH30034.02
21.29 GBP valid until 2024-12-31
Use promo code "23281" to get your promotional price.
Code : A0
Additional Details : Weight : 0.50000kg
Description
- Format: Liquid
Specifications
Store frozen | |
29.2 mg/mL | |
0.1 μm Sterile-filtered |
L-Glutamine, 200mM Solution (29.2mg/mL-Glutamine in 0.85% NaCl), 500mL | |
Liquid | |
500mL |
Safety and Handling
Hazard Statement
Supplemental information
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN AQUATIC-Contains 3% of components with unknown hazards to the aquatic environment.