missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15440809
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reactant in the synthesis of rhenium-glutamine conjugates. It is mainly used in food and pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of rhenium-glutamine conjugates. It is mainly used in food and pharma industry.

Solubility
Soluble 0.3gram in 2ml water clear solution.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39741-62-3
Molecular Formula C9H19ClN2O3
Molecular Weight (g/mol) 238.71
MDL Number MFCD00039081
InChI Key ZEPNUNKDKQACNC-UHFFFAOYNA-N
Synonym h-gln-otbu.hcl, l-glutamine t-butyl ester hydrochloride, h-gln-otbu hcl, s-tert-butyl 2,5-diamino-5-oxopentanoate hydrochloride, h-gln-otbu hydrochloride, l-glutamine tert-butyl ester hydrochloride, hggln-otbu hcl, gln-otbu.hcl, h-gln-otbucl, glutamine t-butyl ester hydrochloride
PubChem CID 15602445
SMILES Cl.CC(C)(C)OC(=O)C(N)CCC(N)=O

Specifications

Color White
Solubility Information Soluble 0.3gram in 2ml water clear solution.
IUPAC Name tert-butyl (2S)-2,5-diamino-5-oxopentanoate;hydrochloride
Quantity 1 g
Formula Weight 238.71
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Glutamine tert-butyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'