missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10214683
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 185°C
Ammonium 0.1% max.
CAS 56-85-9
Arsenic (As) 1ppm max.
pH 4.0 to 6.0
Color White
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 98.5 to 101.0 %
Ignition Residue 0.1% max.
Identification Pass Test
Molecular Formula C5H10N2O3
MDL Number MFCD00008044
Loss on Drying 0.2% max.
Merck Index 15, 4507
Other Amino Acids 1.0% max.
Packaging Glass Bottle
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILES N[C@@H](CCC(N)=O)C(O)=O
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
Specific Rotation 6.3 to 7.3° (+ or -)
Molecular Weight (g/mol) 146.15
PubChem CID 5961
ChEBI CHEBI:18050
Formula Weight 146.15
State of Solution (%T) 95.0% min.
Percent Purity 98.5 to 101.0%
Alpha Vector Glutamine
Quantity 100 g
Physical Form Solid
Chemical Name or Material L-Glutamine
Show More Show Less
Product Identifier
  • L-Glutamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.