missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Histidine, 98% Product Code.: 10665602

Thermo Scientific Chemicals L-Histidine, 98%

Product Code. 10665602
25 g, Plastic Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10665602

Brand: Thermo Scientific Chemicals 166150250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Melting Point 282.0°C
Ammonium 200ppm max.
pH 7.0 to 8.5 (2% aq. soln.)
Color White or Yellow
Chloride 200ppm max.
Chlorine (Cl) 200ppm max.
Foreign Amino Acids 0.5% max. (TLC)
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 98%
Loss on Drying 0.2% max. (105°C, 3 hrs)
Packaging Plastic Bottle
Beilstein 25, 513
Specific Rotation + 12.40
Sulfate 200ppm max.
Merck Index 15, 4758
Transmittance 98% min. (c=5, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 41.6g/L (25 c). Other solubilities: soluble in mineral acids, insoluble in organic solvents
Specific Rotation Condition +12.40° (20°C c=11,6N HCl)
Quantity 25 g
Formula Weight 155.16
Percent Purity 98%
Physical Form Powder, Lumpy Powder or Melt
Chemical Name or Material L-Histidine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0438154 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number166150250,166155000,166151000
Lot NumberA0460517 Certificate TypeCertificate of Analysis Date25/04/2024 Catalog Number166150250,166155000,166151000
Lot NumberA0450782 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number166150250,166155000,166151000
Lot NumberA0451776 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number166150250,166155000,166151000
Lot NumberA0397178 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number166150250,166155000,166151000
Product Identifier
  • L-Histidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Histidine, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.