missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Histidine, 98+% Product Code.: 11418613

Thermo Scientific Chemicals L-Histidine, 98+%

Product Code. 11418613
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11418613

Brand: Thermo Scientific Chemicals A10413.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine acts as a precursor to histamine and a component of carnosine. It is also used in medicine, feed additive, biochemical research, dietary supplement. It is used in a nutrition enhancer, is the amino acid fluid infusion. It can be used in the treatment of gastric ulcer.

Solubility
Soluble in water (15 mg/ml at 20°C).

Notes
Store at 4°C.Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Melting Point ∽281°C (decomposition)
Assay Percent Range >98%
Beilstein 4673585
Merck Index 14,4720
Solubility Information Soluble in water (15mg/ml at 20°C).
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Quantity 25 g
Formula Weight 155.16
Percent Purity ≥98%
Chemical Name or Material L-Histidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252092 Certificate TypeCertificate of Analysis Date26/08/2024 Catalog NumberA10413.14,A10413.36,A10413.22
Lot Number10250080 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog NumberA10413.14,A10413.36,A10413.22
Lot Number10243038 Certificate TypeCertificate of Analysis Date25/08/2022 Catalog NumberA10413.14,A10413.36,A10413.22
Lot Number10238285 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10413.14,A10413.36,A10413.22
Lot Number10224876 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10413.14,A10413.36,A10413.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Histidine, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.