missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Histidine, free base, ≥99%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Specifications
Specifications
Color | White |
Assay Percent Range | ≥99% |
IUPAC Name | (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid |
Quantity | 25 g |
Formula Weight | 155.16 |
Physical Form | Crystalline Powder |
Chemical Name or Material | L-Histidine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Histidine, free base, ≥99%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction