missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Isoleucine, 99%

Product Code. 10771604
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass Bottle
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73-32-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064222 MFCD00004268
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile
PubChem CID 6306
ChEBI CHEBI:17191
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
SMILES CC[C@H](C)[C@H](N)C(O)=O

Specifications

Melting Point 284.0°C to 286.0°C
Color White
Chloride 500ppm max.
Heavy Metals (as Pb) 20ppm max.
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula C2H5CH(CH3)CH(NH2)CO2H
Iron (Fe) 30ppm max.
Beilstein 04, 454
Loss on Drying 0.3% max. (105°C, 3 hrs)
Merck Index 15, 5225
Packaging Plastic Bottle
Solubility Information Solubility in water: 41.2g/L (50°C). Other solubilities: soluble in mineral acids, practically insoluble in alcohol, insoluble in ether
Residue after Ignition 0.3% max.
Specific Rotation + 41.00
Sulfate 300ppm max.
Formula Weight 131.17
Percent Purity 99%
Sublimation Point 168.0 °C
Specific Rotation Condition +41° (20°C c=4,6N HCl)
Quantity 500 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Isoleucine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'