missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Isoleucine, Multi-Compendial, U.S.P., J.T. Baker™
408.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Poly Bottle
Specifications
Packaging | Poly Bottle |
Formula Weight | 131.17 |
Grade | Multi-Compendial/USP |
Quantity | 100 g |
Chemical Name or Material | L-Isoleucine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'