missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lactic acid sodium salt, 99%, extra pure, Thermo Scientific Chemicals
£61.81 - £203.01
Chemical Identifiers
CAS | 867-56-1 |
---|---|
Molecular Formula | C3H5NaO3 |
Molecular Weight (g/mol) | 112.06 |
Synonym | sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11985051
|
Thermo Scientific Acros
439220100 |
10 g | Glass bottle |
£61.81
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11995051
|
Thermo Scientific Acros
439220500 |
50 g | Glass bottle |
£203.01
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
867-56-1 | |
C3H5NaO3 | |
112.06 | |
sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt |
Specifications
161.6°C to 165°C | |
White to Yellow | |
99% | |
CH3CH(OH)CO2Na | |
112.06 | |
99% | |
Authentic |
867-56-1 | |
Powder | |
C3H5NaO3 | |
sodium l-lactate, sodium-l-lactate, sodium lactate, l, sodium s-2-hydroxypropanoate, unii-p2y1c6m9ps, sodium s-lactate, sodium l-+-lactate, p2y1c6m9ps, propanoic acid, 2-hydroxy-, monosodium salt, 2s, l-+-lactic acid sodium salt | |
112.06 | |
Extra Pure | |
L-Lactic acid sodium salt, Extra Pure |
Safety and Handling
missing translation for 'einecsNumber' : 212-762-3
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Lactic acid sodium salt, 99%, extra pure, Thermo Scientific Chemicals