missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Leucine 7-amido-4-methylcoumarin hydrate
Click to view available options
Quantity:
250 mg
Description
A sensitive fluorogenic substrate for leucine aminopeptidase and aminopeptidase M
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 66447-31-2 |
Color | White to Yellow |
Molecular Formula | C16H20N2O3 |
MDL Number | MFCD04966490 |
Synonym | h-leu-amc, 7-leucylamido-4-methylcoumarin, 2s-2-amino-4-methyl-n-4-methyl-2-oxochromen-7-yl pentanamide, l-leucine 7-amido-4-methylcoumarin hydrate, s-2-amino-4-methyl-n-4-methyl-2-oxo-2h-chromen-7-yl pentanamide, l-leu-amc, l-l-amc, leu-mca, ambotzhaa1181, h-leu-mca free base |
InChI Key | GTAAIHRZANUVJS-ZDUSSCGKSA-N |
SMILES | CC(C)C[C@H](N)C(=O)NC1=CC=C2C(C)=CC(=O)OC2=C1 |
IUPAC Name | (2S)-2-amino-4-methyl-N-(4-methyl-2-oxochromen-7-yl)pentanamide |
Molecular Weight (g/mol) | 288.35 |
PubChem CID | 171787 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'