Learn More
Thermo Scientific Chemicals L-Lysine, 98%
Description
L-Lysine is an essential amino acid used in human nutrition. It plays a vital role in calcium absorption, building muscle protein and recovering from surgery or sports injuries. It is used for the treatment of herpes infections and cold sores. Its derivatives lysine acetylsalicylate is utilized to treat pain as well as to detoxify the body after heroin use. It is an important additive to animal feed. Further, it is used in many foods, especially red meats, fish and dairy products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Lysine is an essential amino acid used in human nutrition. It plays a vital role in calcium absorption, building muscle protein and recovering from surgery or sports injuries. It is used for the treatment of herpes infections and cold sores. Its derivatives lysine acetylsalicylate is utilized to treat pain as well as to detoxify the body after heroin use. It is an important additive to animal feed. Further, it is used in many foods, especially red meats, fish and dairy products.
Solubility
Soluble in water. Insoluble in ethanol, ethyl ether, acetone, benzene and common neutral solvent.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 215°C |
Color | White |
Beilstein | 1722531 |
Merck Index | 14,5636 |
Solubility Information | Soluble in water; Insoluble in ethanol,ethyl ether,acetone,benzene and common neutral solvent. |
IUPAC Name | (2S)-2,6-diaminohexanoic acid |
Quantity | 25 g |
Formula Weight | 146.19 |
Optical Rotation | +26° (c=2 in 6M HCl) |
Percent Purity | 98% |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P07L041 | Certificate of Analysis | 10/03/2025 | J62225.22,J62225.36,J62225.14 |
Y21K036 | Certificate of Analysis | 27/01/2025 | J62225.22,J62225.36,J62225.14 |
T15K014 | Certificate of Analysis | 29/07/2024 | J62225.22,J62225.14,J62225.36 |
Q23K029 | Certificate of Analysis | 11/06/2024 | J62225.22,J62225.14,J62225.36 |
P15K006 | Certificate of Analysis | 21/03/2024 | J62225.22,J62225.14,J62225.36 |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
Days
41Hours
14Minutes
35Seconds
34New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.