missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Lysine, 98%
£73.87 - £254.27
Chemical Identifiers
CAS | 56-87-1 |
---|---|
Molecular Formula | C6H14N2O2 |
Molecular Weight (g/mol) | 146.19 |
MDL Number | MFCD00064433 |
InChI Key | KDXKERNSBIXSRK-MLHKIVSYNA-N |
Synonym | l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum |
PubChem CID | 5962 |
ChEBI | CHEBI:18019 |
IUPAC Name | (2S)-2,6-diaminohexanoic acid |
SMILES | NCCCC[C@H](N)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10016203
|
Thermo Scientific Chemicals
303341000 |
100 g | Glass Bottle |
£73.87
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15197412
|
Thermo Scientific Chemicals
303345000 |
500 g | Glass Bottle |
£254.27
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
56-87-1 | |
146.19 | |
KDXKERNSBIXSRK-MLHKIVSYNA-N | |
5962 | |
(2S)-2,6-diaminohexanoic acid |
C6H14N2O2 | |
MFCD00064433 | |
l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum | |
CHEBI:18019 | |
NCCCC[C@H](N)C(O)=O |
Specifications
214.0°C to 216.0°C | |
White to Yellow | |
98% | |
MFCD00064433 | |
l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum | |
KDXKERNSBIXSRK-MLHKIVSYNA-N | |
(2S)-2,6-diaminohexanoic acid | |
5962 | |
146.19 | |
Powder or Crystals |
56-87-1 | |
Authentic | |
C6H14N2O2 | |
+23.0° to +27.0° (20°C, 589nm) (C=2, 1N HCl) | |
3% max. | |
NCCCC[C@H](N)C(O)=O | |
146.19 | |
CHEBI:18019 | |
98% | |
L-Lysine |
Safety and Handling
Product Identifier
- L-Lysine
RUO – Research Use Only