missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Lysine methyl ester dihydrochloride, 99%
Click to view available options
Quantity:
25 g
100 g
500 g
Description
L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
L-Lysine methyl ester dihydrochloride is used in the production of lysine methyl ester based cationic surfactants and hydrogels.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼200°C (decomposition) |
Beilstein | 5302183 |
Solubility Information | Soluble in water. |
Formula Weight | 233.14 |
Percent Purity | 99% |
Quantity | 500 g |
Sensitivity | Hygroscopic |
Chemical Name or Material | L-Lysine methyl ester dihydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'