missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Malic acid, ≥99%, Thermo Scientific Chemicals
Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
Specifications
Specifications
Melting Point | 100°C to 106°C |
Beilstein | 1723541 |
Formula Weight | 134.09 |
Percent Purity | ≥99% |
Density | 1.6 |
Quantity | 500 g |
Flash Point | >110°C |
Packaging | Plastic bottle |
Physical Form | Crystalline Powder |
Specific Gravity | 1.6 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
L-Malic acid, ≥99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction