Learn More
L(-)-Malic acid, 99%, Thermo Scientific Chemicals
£19.10 - £132.00
Chemical Identifiers
CAS | 97-67-6 |
---|---|
Molecular Formula | C4H6O5 |
Molecular Weight (g/mol) | 134.087 |
MDL Number | MFCD00064213 |
InChI Key | BJEPYKJPYRNKOW-REOHCLBHSA-N |
Synonym | l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid |
PubChem CID | 222656 |
ChEBI | CHEBI:30797 |
IUPAC Name | (2S)-2-hydroxybutanedioic acid |
SMILES | C(C(C(=O)O)O)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10113010
|
Thermo Scientific Acros
150590250 |
25 g | Plastic bottle |
£19.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10295290
|
Thermo Scientific Acros
150591000 |
100 g | Plastic bottle |
£41.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10637744
|
Thermo Scientific Acros
150595000 |
500 g | Plastic bottle |
£132.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
97-67-6 | |
134.087 | |
BJEPYKJPYRNKOW-REOHCLBHSA-N | |
222656 | |
(2S)-2-hydroxybutanedioic acid |
C4H6O5 | |
MFCD00064213 | |
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid | |
CHEBI:30797 | |
C(C(C(=O)O)O)C(=O)O |
Spezifikation
97-67-6 | |
White | |
C4H6O5 | |
MFCD00064213 | |
15, 5771 | |
BJEPYKJPYRNKOW-REOHCLBHSA-N | |
(2S)-2-hydroxybutanedioic acid | |
222656 | |
134.09 | |
Authentic | |
L(-)-Malic acid |
99°C to 107°C | |
99% | |
HO2CCH2CH(OH)CO2H | |
03, 419 | |
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid | |
C(C(C(=O)O)O)C(=O)O | |
134.087 | |
CHEBI:30797 | |
≥98.5% | |
Crystalline Powder or Granules |
Sicherheit und Handhabung
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only