missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Menthone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
Description
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.
Solubility
Soluble in ether, alcohol, benzene, acetone.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.
Solubility
Soluble in ether, alcohol, benzene, acetone.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
Density | 0.893 |
Boiling Point | 208°C to 210°C |
Flash Point | 72°C (162°F) |
Assay Percent Range | 97% |
Refractive Index | 1.45 |
Quantity | 50 g |
Beilstein | 3648743 |
Merck Index | 14,5838 |
Solubility Information | Soluble in ether,alcohol,benzene,acetone. |
Formula Weight | 154.25 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'