missing translation for 'onlineSavingsMsg'
Learn More

L-Menthone, 97%, Thermo Scientific Chemicals

Product Code. 11474644
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

Solubility
Soluble in ether, alcohol, benzene, acetone.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14073-97-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001634,MFCD00136033
InChI Key NFLGAXVYCFJBMK-BDAKNGLRSA-N
Synonym --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one
PubChem CID 26447
ChEBI CHEBI:15410
IUPAC Name (2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one
SMILES CC(C)[C@@H]1CC[C@@H](C)CC1=O

Specifications

Density 0.893
Boiling Point 208°C to 210°C
Flash Point 72°C (162°F)
Assay Percent Range 97%
Refractive Index 1.45
Quantity 250 g
Beilstein 3648743
Merck Index 14,5838
Solubility Information Soluble in ether,alcohol,benzene,acetone.
Optical Rotation −25° (Neat)
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material L-Menthone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'