missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Methionine, 98+% Product Code.: 10685602

Thermo Scientific Chemicals L-Methionine, 98+%

Product Code. 10685602
25 g, Plastic Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10685602

Brand: Thermo Scientific Chemicals 166160250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilaninShow MoreShow Less
PubChem CID 6137
ChEBI CHEBI:16643
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point 276.0°C to 279.0°C
Ammonium 300ppm max.
pH 5.5 to 6.5 (aq. soln. 1 in 100)
Color White
Chloride 500ppm max.
Heavy Metals (as Pb) 15ppm max.
Individual Impurity 0.5% max. of any individual impurity
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 98+%
Loss on Drying 0.5% max. (1 g, 105°C)
Packaging Plastic Bottle
Linear Formula CH3SCH2CH2CH(NH2)CO2H
Beilstein 04, II, 938
Specific Rotation + 23.25
Sulfate 300ppm max.
Merck Index 15, 6047
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: soluble in water: 53.7g/L (20°C). Other solubilities: insoluble in abs.alcohol, ether, petr.ether,, benzene,acetone, ethanol, diethylether, soluble in mineral acids, soluble in warm, dilute alcohol
IUPAC Name (2S)-2-amino-4-(methylsulfanyl)butanoic acid
Specific Rotation Condition +23.25° (20°C c=2,6N HCl)
Quantity 25 g
Formula Weight 149.21
Percent Purity ≥98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Methionine, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464758 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number166161000,166160250,166165000,166160025
Lot NumberA0449931 Certificate TypeCertificate of Analysis Date27/02/2023 Catalog Number166161000,166160250,166165000,166160025
Lot NumberA0396526 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number166161000,166160250,166165000,166160025
Lot NumberA0403300 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number166161000,166160250,166165000,166160025
Lot NumberA0409768 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number166161000,166160250,166165000,166160025
Product Identifier
  • L-Methionine
  • Multiple Manufacturers
    Explore our new promotion and get up to 70% off some of our most popular products.
    Promo Code: 24127

    Expires2025-05-31

  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24155

    Expires2025-04-30

    Hot Deal

    *Discount will apply to qualifying orders received by Fisher Scientific between 01/03/2025 and 31/03/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.

    Terms & Conditions See Full Promo Details

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Methionine, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.