missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine, 98+%

Product Code. 10689853
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-68-3
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00063097 MFCD00801344
InChI Key FFEARJCKVFRZRR-BYPYZUCNSA-N
Synonym l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
PubChem CID 6137
ChEBI CHEBI:16643
SMILES CSCC[C@H](N)C(O)=O

Specifications

Melting Point 276.0°C to 279.0°C
Ammonium 300ppm max.
pH 5.5 to 6.5 (aq. soln. 1 in 100)
Color White
Chloride 500ppm max.
Heavy Metals (as Pb) 15ppm max.
Individual Impurity 0.5% max. of any individual impurity
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 98+%
Loss on Drying 0.5% max. (1 g, 105°C)
Packaging Plastic Bottle
Linear Formula CH3SCH2CH2CH(NH2)CO2H
Beilstein 04, II, 938
Specific Rotation + 23.25
Sulfate 300ppm max.
Merck Index 15, 6047
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: soluble in water: 53.7g/L (20°C). Other solubilities: insoluble in abs.alcohol, ether, petr.ether,, benzene,acetone, ethanol, diethylether, soluble in mineral acids, soluble in warm, dilute alcohol
IUPAC Name (2S)-2-amino-4-methylsulfanylbutanoic acid
Specific Rotation Condition +23.25° (20°C c=2,6N HCl)
Quantity 2.5 kg
Formula Weight 149.21
Percent Purity ≥98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Methionine, 98+%
Show More Show Less
Product Identifier
  • L-Methionine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.