missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine, 98+%

Catalog Number p-7080685
Click to view available options
Quantity:
100 g
500 g
2500 g
63-68-3
C5H11NO2S
149.21
MFCD00063097 MFCD00801344
FFEARJCKVFRZRR-BYPYZUCNSA-N
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
6137
CHEBI:16643
(2S)-2-amino-4-methylsulfanylbutanoic acid
CSCC[C@H](N)C(O)=O
63-68-3
C5H11NO2S
149.21
MFCD00063097 MFCD00801344
FFEARJCKVFRZRR-BYPYZUCNSA-N
l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin
6137
CHEBI:16643
(2S)-2-amino-4-methylsulfanylbutanoic acid
CSCC[C@H](N)C(O)=O

L-Methionine (CAS # 63-68-3) is an essential amino acid that contains sulphur. L-Methionine is involved in cellular detoxification in several species.

L-Methionine iserves as a chelating agent for heavy metals, as a flavoring agent and nutritional supplement for foods. It acts as a feed additive, vegetable oil enrichment and as a single cell protein. In addition to this, it is used as a hepatoprotectant and also acts as a lipotropic agent and prevents excess fat buildup in the liver.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• L-Methionine is an essential amino acid required for tissue repair and maintenance

• L-Methionine affects absorption of selenium and zinc

• L-Methionine can assist cellular detoxification by chelating lead and mercury

• L-Methionine exhibitis properties associated with anti-aging and can also prevent the buildup of fat in the liver

Application

• L-Methionine is appropriate for use in cell culture and biochemistry laboratory applications

• It is useful for the laboratory study of methionine pathways in the cell relating to aging, degeneration, and metabolism

TRUSTED_SUSTAINABILITY
Melting Point ∼280°C (decomposition)
Beilstein 1722294
Merck Index 14,5975
Solubility Information Soluble in water and hydrochloride. Slightly soluble in acetic acid. Insoluble in ethanol,benzene,acetone and ether.
Formula Weight 149.21
Percent Purity ≥98%
Quantity 100 g
Optical Rotation +23° (c=2 in 2N HCl)
Chemical Name or Material L-Methionine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'