Learn More
Gibco™ L-Methionine (Methyl-13C)
L-Methionine is a common sulfur-containing amino acid that is commonly used for the isotopic labeling of methionine protein residues for NMR spectroscopy studies.
Brand:  Gibco™ A39248
Additional Details : Weight : 0.01430kg
Product Code. 16525651
  • £209.00 / 225mg
Estimated Shipment: 19-07-2024
to see stock.
Description
Description
Methionine is a common methyl-group donor to various substrates, such as creatine, epinephrine, ergosterol, and choline.
EC Number: 200-562-9 (unlabeled)
Specifications
Specifications
(S)-2-Amino-4-(Methylmercapto)butyric acid / L-2-Amino-4-(methylthio)cutanoic acid
>98%
Powder
150.2
225 mg
Wet Ice