missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Nicotine, 99+%
35.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- L-Nicotine is a naturally occuring alkaloid in the nightshade family of plants
- It is also known as 3-(1-methyl-2-pyrrolidinyl)pyridine or (S)-(-)-nicotine
Applications
- L-Nicotine has been shown in studies to alleviate smoking habits
- It has also been shown to enhance cognitive functions such as memory
- Studies show that lower levels of nicotine in the form of gum may also help eliminate smoking habits
Specifications
Melting Point | -80.0°C |
Color | Brown or Colorless to Yellow |
Density | 1.0150g/mL |
Boiling Point | 247.0°C |
Flash Point | 101°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.5255 to 1.5275 |
Quantity | 25 g |
Show More |
Safety and Handling
Product Identifier
- L-Nicotine
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H300-Fatal if swallowed.
- H310-Fatal in contact with skin.
- H330-Fatal if inhaled.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P361-Take off immediately all contaminated clothing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P310-Immediately call a POISON CENTER/doctor/.
- P330-Rinse mouth.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'