missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Ornithine hydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.
Solubility
Soluble in water (543 mg/ml at 20°C), and alcohol.
Notes
Store at room temperature. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.
Solubility
Soluble in water (543 mg/ml at 20°C), and alcohol.
Notes
Store at room temperature. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
Specifications
Specifications
Melting Point | ∼238°C (decomposition) |
Beilstein | 3625847 |
Merck Index | 14,6874 |
Solubility Information | Soluble in water (543mg/ml at 20°C),and alcohol. |
Quantity | 100 g |
Formula Weight | 168.62 |
Optical Rotation | +23° (c=5 in 5N HCl) |
Percent Purity | 99% |
Chemical Name or Material | L-Ornithine hydrochloride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction