missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 11450043
Click to view available options
Quantity:
25 g
100 g
500 g

L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Ornithine hydrochloride is used in the biosynthesis of L-arginine, L-proline and polyamines. It plays an important role in ammonia metabolism via the urea cycle.

Solubility
Soluble in water (543 mg/ml at 20°C), and alcohol.

Notes
Store at room temperature. Store in cool, dry place in a tightly sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.621
MDL Number MFCD00064562
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
PubChem CID 76654
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Melting Point ∼238°C (decomposition)
Beilstein 3625847
Merck Index 14,6874
Solubility Information Soluble in water (543mg/ml at 20°C),and alcohol.
Quantity 500 g
Formula Weight 168.62
Optical Rotation +23° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material L-Ornithine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.