missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine methyl ester dihydrochloride, 97%, Thermo Scientific Chemicals

£24.00 - £286.00

Chemical Identifiers

CAS 40216-82-8
Molecular Formula C6H16Cl2N2O2
Molecular Weight (g/mol) 219.106
MDL Number MFCD00190977
InChI Key SNFZNVWAQDVFAK-XRIGFGBMSA-N
PubChem CID 14273060
IUPAC Name methyl (2S)-2,5-diaminopentanoate;dihydrochloride
SMILES COC(=O)C(CCCN)N.Cl.Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15419808
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63225.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15429808
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63225.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15439808
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H63225.22
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

40216-82-8
219.106
SNFZNVWAQDVFAK-XRIGFGBMSA-N
methyl (2S)-2,5-diaminopentanoate;dihydrochloride
C6H16Cl2N2O2
MFCD00190977
14273060
COC(=O)C(CCCN)N.Cl.Cl

Specifications

40216-82-8
C6H16Cl2N2O2
h-orn-ome.2hcl, l-ornithine methyl ester dihydrochloride, methyl l-ornithine dihydrochloride, s-methyl 2,5-diaminopentanoate dihydrochloride, h-orn-ome?hcl, methyl l-ornithine hcl, h-orn-ome 2hcl, h-orn-ome2hcl, l-ornithine, methyl ester, dihydrochloride, h-orn-ome 2 hcl
COC(=O)C(CCCN)N.Cl.Cl
219.106
219.11
Powder
White
MFCD00190977
SNFZNVWAQDVFAK-XRIGFGBMSA-N
methyl (2S)-2,5-diaminopentanoate;dihydrochloride
14273060
97%
L-Ornithine methyl ester dihydrochloride
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 254-841-5

missing translation for 'tsca' : No

missing translation for 'storageNote' : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only