missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine, 98.5-101.0%, Thermo Scientific Chemicals

Product Code. 15279846
Click to view available options
Quantity:
25 g
100 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-oh
PubChem CID 6140
ChEBI CHEBI:17295
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 270.0°C to 275.0°C
pH 5.4 to 6.0 (1% solution)
Chloride 0.05% max.
Heavy Metals 15ppm max.
Infrared Spectrum Authentic
Linear Formula C6H5CH2CH(NH2)CO2H
Iron (Fe) 0.003% max.
Loss on Drying 0.3% max. (105°C, 3 hrs)
Solubility Information Solubility in water: 27g/L (20°C). Other solubilities: very slightly soluble in methanol, ethanol
IUPAC Name (2S)-2-amino-3-phenylpropanoic acid
Specific Rotation −34.10
Sulfate 0.03% max.
Formula Weight 165.19
Sulfated Ash 0.4% max.
Specific Rotation Condition −34.10 (20.00°C c=2,water,dry basis)
Quantity 1 kg
Chemical Name or Material L-Phenylalanine, 98.5-101.0%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'