missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L-Phenylalanine, 98.5+% Product Code.: 10574061

Thermo Scientific Chemicals L-Phenylalanine, 98.5+%

Product Code. 10574061
25 g, Plastic Bottle
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy

Product Code. 10574061

Brand: Thermo Scientific Chemicals 130310250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-91-2
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00064227
InChI Key COLNVLDHVKWLRT-UHFFFAOYNA-N
Synonym l-phenylalanine, phenylalanine, s-2-amino-3-phenylpropanoic acid, 3-phenyl-l-alanine, s-phenylalanine, 3-phenylalanine, 2s-2-amino-3-phenylpropanoic acid, s-2-amino-3-phenylpropionic acid, beta-phenyl-l-alanine, h-phe-ohShow MoreShow Less
PubChem CID 6140
ChEBI CHEBI:17295
SMILES NC(CC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 270.0°C to 275.0°C
pH 5.4 to 6.0 (1% aq. soln.)
Color White
Heavy Metals (as Pb) 20ppm max.
Infrared Spectrum Authentic
Lead (Pb) 10ppm max.
Assay Percent Range 98.5+%
Loss on Drying 0.3% max. (105°C, 3 hrs)
Packaging Plastic Bottle
Linear Formula C6H5CH2CH(NH2)CO2H
Specific Rotation − 34.10
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 27g/L (20°C). Other solubilities: very slightly soluble in methanol, ethanol
IUPAC Name 2-amino-3-phenylpropanoic acid
Specific Rotation Condition − 34.10 (20.00°C c=2,water,dry basis)
Quantity 25 g
Formula Weight 165.19
Percent Purity ≥98.5%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Phenylalanine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0452521 Certificate TypeCertificate of Analysis Date26/04/2023 Catalog Number130315000,130310250,130311000,130310025
Lot NumberA0450262 Certificate TypeCertificate of Analysis Date10/02/2023 Catalog Number130315000,130310250,130311000,130310025
Lot NumberA0401146 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number130315000,130310250,130311000,130310025
Lot NumberA0410852 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number130315000,130310250,130311000,130310025
Lot NumberA0407468 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number130315000,130310250,130311000,130310025

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L-Phenylalanine, 98.5+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.