missing translation for 'onlineSavingsMsg'
Learn More
L-Phenylalaninol, 98%, Thermo Scientific Chemicals Product Code.: 11494653

L-Phenylalaninol, 98%, Thermo Scientific Chemicals

Product Code. 11494653
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11494653

Brand: Thermo Scientific Alfa Aesar A11586.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

L-Phenylalaninol acts as an antiulcer agent that reduces the secretion of gastric acid and prevents ulcer formation. It inhibits intestinal absorption of L-phenylalanine making it a prospective treatment for phenylketonuria. Further, it reacts with nitriles to form oxazolines, which finds application in palladium-catalyzed allylic substitution. In addition to this, it is also used in amidation for chiral resolution and nicotinamide adenine dinucleotide modeling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Phenylalaninol acts as an antiulcer agent that reduces the secretion of gastric acid and prevents ulcer formation. It inhibits intestinal absorption of L-phenylalanine making it a prospective treatment for phenylketonuria. Further, it reacts with nitriles to form oxazolines, which finds application in palladium-catalyzed allylic substitution. In addition to this, it is also used in amidation for chiral resolution and nicotinamide adenine dinucleotide modeling.

Solubility
Soluble in chloroform, ethyl acetate, ethanol and methanol.

Notes
Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3182-95-4
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00004732
InChI Key STVVMTBJNDTZBF-VIFPVBQESA-N
Synonym l-phenylalaninol, s-2-amino-3-phenyl-1-propanol, s---2-amino-3-phenyl-1-propanol, s-2-amino-3-phenylpropan-1-ol, h-phenylalaninol, h-phe-ol, 2s-2-amino-3-phenylpropan-1-ol, phenylalaninol, --l-phenylalaninol, l-phenyl alaninolShow MoreShow Less
PubChem CID 447213
IUPAC Name (2S)-2-amino-3-phenylpropan-1-ol
SMILES N[C@H](CO)CC1=CC=CC=C1

Specifications

Melting Point 89°C to 94°C
Quantity 5 g
Optical Rotation −23° (c=5 in ethanol)
Sensitivity Air Sensitive
UN Number UN3259
Beilstein 2208238
Solubility Information Soluble in chloroform,ethyl acetate,ethanol and methanol.
Formula Weight 151.21
Percent Purity 98%
Chemical Name or Material L-Phenylalaninol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10250499 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA11586.03,A11586.14,A11586.06
Lot Number10246274 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA11586.03,A11586.14,A11586.06
Lot Number10245217 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog NumberA11586.03,A11586.14,A11586.06
Lot NumberAI3653A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11586.03,A11586.14,A11586.06
Lot Number10215958 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11586.03,A11586.14,A11586.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
L-Phenylalaninol, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.