Learn More
Thermo Scientific Chemicals L(-)-Proline, 99+%
L-Proline CAS 147-85-3, (synonym: (S)-Pyrrolidine-2-carboxylic acid) is a nonessential amino acid and building block of proteins that has applications such as acting as catalyst in chemical reactions.
Chemical Identifiers
CAS | 147-85-3 |
---|---|
Molecular Formula | C5H9NO2 |
Molecular Weight (g/mol) | 115.13 |
MDL Number | MFCD00064318 |
InChI Key | ONIBWKKTOPOVIA-UHFFFAOYNA-N |
Synonym | l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh |
PubChem CID | 145742 |
ChEBI | CHEBI:17203 |
SMILES | OC(=O)C1CCCN1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• L-Proline is an amino acid that is found as a component of various proteins.
Application
• L-Proline is used as asymmetric catalyst in organic synthesis and asymmetric aldol cyclization
• It is also used along with ninhydrin in chromatography
• It is involved in the Michael addition of dimethyl malonate to alfa-beta-unsaturated aldehydes
• It is used in pharmaceutical and biotechnological applications due to its osmoprotactant property
• It is precursor of hydroxyproline in collage and involved in proper functioning of joint and tendons
Chemical Identifiers
147-85-3 | |
115.13 | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
145742 | |
OC(=O)C1CCCN1 |
C5H9NO2 | |
MFCD00064318 | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
CHEBI:17203 |
Specifications
200ppm max. | |
1ppm max. | |
White | |
10ppm max. | |
10ppm max. | |
0.3% max. (105°C, 3 hrs) | |
C5H9NO2 | |
22, 2 | |
06,492; 08,421; 09,393; 10,331; 11,446; 13,292 | |
15, 7895 | |
0.1% max. | |
ONIBWKKTOPOVIA-UHFFFAOYNA-N | |
OC(=O)C1CCCN1 | |
− 85.50 (20.00°C c=4,H2O) | |
145742 | |
115.13 | |
Crystalline Powder or Crystals |
147-85-3 | |
5.5 to 7 (2.5% soln. at 25°C) | |
200ppm max. | |
Authentic | |
99+% | |
0.5% max. (TLC) | |
MFCD00064318 | |
-84.5° to -86.8° (20°C, 589nm) (c=4, H2O) | |
200ppm max. | |
95% min. (c=10, H2O, 430nm) 1cm cell | |
l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh | |
1.3500g/mL | |
(2S)-pyrrolidine-2-carboxylic acid | |
115.13 | |
CHEBI:17203 | |
99.0 to 101.0% | |
L(-)-Proline |
RUO – Research Use Only