missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Proline, Multi-Compendial, U.S.P., J.T. Baker™
2327.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
1 kg
Packaging:
Poly Bottle
Specifications
CAS | 147-85-3 |
Molecular Formula | C5H9NO2 |
MDL Number | MFCD00064318 |
Packaging | Poly Bottle |
Synonym | l-proline, proline, l---proline, s-pyrrolidine-2-carboxylic acid, 2-pyrrolidinecarboxylic acid, --proline, --s-proline, 2s-pyrrolidine-2-carboxylic acid, prolinum, h-pro-oh |
InChI Key | ONIBWKKTOPOVIA-UHFFFAOYNA-N |
SMILES | OC(=O)C1CCCN1 |
IUPAC Name | (2S)-pyrrolidine-2-carboxylic acid |
Molecular Weight (g/mol) | 115.13 |
PubChem CID | 145742 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'