missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals L(+)-Rhamnose monohydrate, 99% Product Code.: 10464785

Thermo Scientific Chemicals L(+)-Rhamnose monohydrate, 99%

Product Code. 10464785
500 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10464785

Brand: Thermo Scientific Chemicals 174085000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10030-85-0
Molecular Formula C6H12O5
Molecular Weight (g/mol) 164.16
MDL Number MFCD00149363,MFCD00136036
InChI Key SHZGCJCMOBCMKK-HGVZOGFYSA-N
Synonym l-+-rhamnose monohydrate, 2r,3r,4s,5s-2,3,4,5-tetrahydroxyhexanal hydrate, l + rhamnopyranose, l-mannose, 6-deoxy-, monohydrate, 6-deoxy-l-mannose hydrate, rhamnose hydrate, l-rha hydrate, l-rhamnose hydrate, l +-rhamnose hydrate, a-l-rhamnose monohydrate
PubChem CID 20849066
IUPAC Name (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate
SMILES C[C@@H]1O[C@@H](O)[C@H](O)[C@H](O)[C@H]1O

Specifications

Melting Point 91.0°C to 93.0°C
Color White
Ignition Residue 0.2% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 500 g
Specific Rotation Condition + 8.20 (20.00°C c=4.5,H2O)
Specific Rotation + 8.20
Solubility Information Solubility in water: 300g/L (20°C). Other solubilities: soluble in ethanol and methanol
Formula Weight 182.17
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Rhamnose monohydrate
Show More Show Less
Product Identifier
  • L(+)-Rhamnose monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals L(+)-Rhamnose monohydrate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.