missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(+)-Rhamnose, monohydrate, ∽99%, MP Biomedicals™
Click to view available options
Quantity:
100 g
Specifications
Color | White |
Density | 1.47g/mL |
Assay Percent Range | ∽ 99% |
Quantity | 100 g |
IUPAC Name | (2R,3R,4S,5S)-2,3,4,5-tetrahydroxyhexanal;hydrate |
Formula Weight | 182.17 |
Percent Purity | ∽99% |
Physical Form | Solid |
Chemical Name or Material | L-(+)-Rhamnose, monohydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'