missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Fixed-Speed Drive

Masterflex™ L/S™ Fixed-Speed Drive intended for reliable, general transfer and feed applications

Brand:  Masterflex™ 07540-30

 View more versions of this product

Product Code. 11713969

  • £694.00 / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Economical drives for reliable general transfer and feed applications
  • Painted Steel housed construction is IP 22 rated
  • Choose from seven speeds based on flow rate or number of heads required
  • Economy fixed-speed drives are stackable and ideal for lab, field, or process feed
  • Compact drives are ideal for applications where space is limited
  • Lighted power switch on the front of the drive indicates when the pump is running
  • Compatible with five different pump head types: Standard, Easy-Load™, Easy-Load™ II, Easy-Load™ 3, and Cartridge Pump Heads
  • Accepts up to 2 pump heads
Specifications

Specifications

0.9 amps
152 x 134 x 118 mm
Standard, Easy-Load, Easy-Load II, Easy-Load 3, Cartridge Pump Heads
Painted Steel
Fixed
Fixed Speed
1.8 to 140 mL/min.
IP 22
2 Pump Heads Accepted
30 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers