missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masterflex™ L/S™ PTFE-Diaphragm Pump System
Masterflex™ L/S™ PTFE-Diaphragm Pump System is ideal for chemical feed, metering applications that require high-purity and/or high-pressure
Brand: Masterflex™ 77915-10
This item is not returnable.
View return policy
Description
- L/S™ precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation
- Large LED display and LED indicators confirm motor rpm and motor direction at a glance
- Complete pump systems include all you need to start pumping
- Ideal for chemical feed and metering applications that require high-purity, and/or high-pressure
- Flow range 80 to 800 mL/min - flow rate depends on drive rpm and tubing size
- PTFE wetted parts help maintain fluid purity
- Pump through filters or into pressurized system
- Check valve on inlet side of pump holds prime for up to two hours when pump is off
- Unique geometry minimizes fluid churning and dead volume for high metering accuracy without reagent contamination
Specifications
Variable Speed Digital, ±0.25% Speed Control | |
6 to 600 rpm | |
(43.94 x 21.08 x 21.59 cm) | |
80 to 800 mL/min. | |
IP33-rated |
1.8 A at 115 V, 1.2 A at 230 V | |
90 to 260 VAC | |
Polytetrafluoroethylene (PTFE) | |
L/S PTFE-Diaphragm Pump Head (07090-42) |