missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ L/S™ Standard Long-Shaft Pump Heads

Masterflex™ L/S™ Standard Long-Shaft Pump Heads are ideal for fluid transfer applications that require high accuracy and repeatability

Brand:  Masterflex™ 07024-10

 View more versions of this product

Product Code. 15316918

  • £485.00 / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Ideal for fluid transfer applications requiring high accuracy and repeatability
  • Standard heads are precision molded to accommodate a single size of tubing
  • Pump head is compact and simple in design
  • Same side tubing entry/exit is perfect for space-limited applications
  • Standard heads are compact and easy to integrate into your OEM applications
  • Suitable for use with L/S™ 16, or 24 tubing size
  • Accepts a single tubing size to ensure optimal performance of Masterflex pumps
  • Use with Masterflex L/S™ pump drives that require the long-shaft style pump head
  • These pump heads are not compatible with standard shaft drives
Specifications

Specifications

1:1 Gear Ratio
Fluid transfer that requires high accuracy and repeatability
Polycarbonate
3
L/S™ 24
Stainless Steel
2.8 to 1700 mL/min.
L/S™ Pump Drives that require a Long-Shaft Pump Head style
1
Fixed
Long-Shaft
0.02 to 600 rpm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers