Learn More
Masterflex™ L/S™ Standard Pump Head Replacement Rotor Assembly
Masterflex™ L/S™ Standard Pump Head Replacement Rotor Assembly extends the operating life of standard pump heads
Brand: Masterflex™ 07015-90
Code : A0
Additional Details : Weight : 0.80000kg
Description
Masterflex™ L/S™ Standard Pump Head Replacement Rotor Assembly extends the operating life of standard pump heads
- Replacement parts extend the operating life of standard pump heads
- Two end bell assemblies are required for a complete head assembly
- Choose replacement parts suitable for your I/P™ standard pump head size
- Suitable for use with L/S™ Standard Pump Heads 07015-21, 07015-52, 07024-21 and 07024-52
Specifications
L/S Standard Pump Heads 07015-21, 07015-52, 07024-21, and 07024-52 | |
Stainless Steel | |
High-Performance Precision Tubing | |
Replacement Rotor Assembly |