Learn More
Invitrogen™ L-selectin (SELL) Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EMSELLX5
Beschreibung
The Mouse L-Selectin (SELL) ELISA quantitates Ms L-Selectin in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms L-Selectin. Principle of the method The Mouse L-Selectin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
CD62L (L-selectin) is an adhesion glycoprotein with a molecular weight range of 74-95 kDa that is constitutively expressed on the cell surface of leukocytes, and mediates their homing to inflammatory sites and peripheral lymph nodes by enabling rolling along the venular wall. CD62L is also involved in activation-induced neutrophil aggregation. Activation-dependent CD62L shedding, however, counteracts neutrophil rolling. CD62L has also signaling roles including enhance of chemokine receptor expression. Similar to CD62P, the major ligand of CD62L is PSGL-1 (P-selectin glycoprotein ligand-1). Single-nucleotide polymorphisms in the CD26L gene have been associated with various diseases including immunoglobulin A nephropathy. Alternatively spliced transcript variants have been found for CD26L.Spezifikation
P18337 | |
35 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
20343 | |
<12% | |
SAV-HRP, Stop Solution, Wash Buffer, Adhesive Plate Covers, Standard, Chromogen, Pre-coated 96 well plates, Biotinylated Detection Antibody, Assay Diluent Concentrate | |
5 x 96 Tests | |
Bone Research | |
2°C to 8°C | |
Mouse | |
4 hr. 45 min. |
40.96 pg/mL to 10,000 pg/mL | |
35 pg/mL to 10,000 pg/mL | |
Biotin | |
Other Proteins | |
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN (OB), LIX, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF. | |
Mouse | |
Colorimetric Microplate Reader | |
AI528707,CD62L,L-selectin,LECAM-1,Lnhr,Ly-22,Ly-m22,Lyam-1,Lyam1 | |
<10% | |
HRP | |
RUO | |
Plasma, 1 μ;L; Serum, 1 μ;L; Supernatant, 100 μ;L | |
AI528707, CD62L, L-selectin, LECAM-1, Lnhr, Ly-22, Ly-m22, Lyam-1, Lyam1 | |
1 hr. 20 min. |