Learn More
Gibco™ L-Selenomethionine
L-Selenomethionine is a naturally occurring amino acid containing selenium that is commonly used for the labeling of proteins to facilitate structural determination by x-ray crystallography.
Brand: Gibco™ A39247
Additional Details : Weight : 0.02990kg
Description
Selenomethionine is an antioxidant and has been shown to increase glutathione peroxidase activity in endothelial cellsSpecifications
(S)-2-Amino-4-(methylseleno)butyric acid | |
MFCD00037210 | |
C[Se]CC[C@H](N)C(O)=O | |
196.12 | |
250 mg | |
Wet Ice |
C5H11NO2Se | |
RJFAYQIBOAGBLC-BYPYZUCNSA-N | |
(2S)-2-amino-4-(methylselanyl)butanoic acid | |
>98% | |
Powder |
For Research Use Only. Not for use in diagnostic procedures.