missing translation for 'onlineSavingsMsg'
Learn More

L-Serinamide hydrochloride, 98%, Thermo Scientific™

Product Code. 10073550
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 65414-74-6
Molecular Formula C3H9ClN2O2
Molecular Weight (g/mol) 140.567
MDL Number MFCD00067560
InChI Key VURWDDZIWBGXCK-DKWTVANSSA-N
Synonym l-serinamide hydrochloride, h-ser-nh2.hcl, l-serine amide hydrochloride, s-2-amino-3-hydroxypropanamide hydrochloride, 2s-2-amino-3-hydroxypropanamide hydrochloride, propanamide, 2-amino-3-hydroxy-, monohydrochloride, 2s, s-2-amino-3-hydroxypropionamide hydrochloride, h-ser-nh hcl, h-ser-nh2cl, h-ser-nh2?cl
PubChem CID 16212403
IUPAC Name (2S)-2-amino-3-hydroxypropanamide;hydrochloride
SMILES C(C(C(=O)N)N)O.Cl

Specifications

Melting Point 184°C to 190°C
Color Purple to Red
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula HOCHCH(NH)CONH·HCl
Packaging Glass Bottle
Specific Rotation +13.5° to +15.5° (20°C, 589nm) (c=1, H2O)
Formula Weight 140.57
Percent Purity ≥97.5% (Argentometry)
Specific Rotation Condition +14.5° (20°C c=1,H2O)
Quantity 1 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Serinamide hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'