missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine, 99%

Product Code. 10205330 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10205330

Brand: Thermo Scientific Chemicals 132660250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 222.0°C
Ammonium 200ppm max.
Arsenic (As) 1ppm max.
pH 5.0 to 6.5 (2.5% soln. at 25°C)
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 0.2% (105°C, 3 hrs) max.
Other Amino Acids 0.5% (TLC) max.
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2H
Specific Rotation 15.2
Sulfate 200ppm max.
Transmittance 95% (c=10, H2O, 430nm) 1cm cellmin.
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene, soluble in mineral acids, insoluble in organic solvents, insoluble in ethanol and diethylether
Sublimation Point 0150.0°C
Density 1.6000g/mL
Specific Rotation Condition 15.20 (20.00°C c=10,2N HCl)
Quantity 25 g
Formula Weight 105.09
Percent Purity 99%
Specific Gravity 1.6
Physical Form Crystalline Powder, Crystals or Crystalline Needles
Chemical Name or Material L-Serine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.