missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine, 99%

Catalog Number p-4509911
36.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
56-45-1
C3H7NO3
105.093
MTCFGRXMJLQNBG-REOHCLBHSA-N
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
5951
CHEBI:17115
(2S)-2-amino-3-hydroxypropanoic acid
C(C(C(=O)O)N)O
56-45-1
C3H7NO3
105.093
MTCFGRXMJLQNBG-REOHCLBHSA-N
l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
5951
CHEBI:17115
(2S)-2-amino-3-hydroxypropanoic acid
C(C(C(=O)O)N)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 222.0°C
Ammonium 200ppm max.
Arsenic (As) 1ppm max.
pH 5.0 to 6.5 (2.5% soln. at 25°C)
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula HOCH2CH(NH2)CO2H
Iron (Fe) 10ppm max.
Loss on Drying 0.2% (105°C, 3 hrs) max.
Other Amino Acids 0.5% (TLC) max.
Packaging Glass Bottle
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: insoluble in alcohol, ether and benzene, soluble in mineral acids, insoluble in organic solvents, insoluble in ethanol and diethylether
Specific Rotation 15.2
Sulfate 200ppm max.
Formula Weight 105.09
Transmittance 95% (c=10, H2O, 430nm) 1cm cellmin.
Percent Purity 99%
Sulfated Ash 0.1% max.
Sublimation Point 0150.0°C
Density 1.6000g/mL
Specific Rotation Condition 15.20 (20.00°C c=10,2N HCl)
Quantity 25 g
Specific Gravity 1.6
Physical Form Crystalline Powder, Crystals or Crystalline Needles
Chemical Name or Material L-Serine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'