missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Serine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.
Solubility
Soluble in water (50 mg/ml), and methanol
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
L-Serine methyl ester hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields. It is also a used as a raw material in peptide synthesis.
Solubility
Soluble in water (50 mg/ml), and methanol
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 155°C (decomposition) |
Beilstein | 3559591 |
Solubility Information | Soluble in water (50mg/ml),and methanol |
Formula Weight | 155.58 |
Percent Purity | 98% |
Quantity | 5 g |
Optical Rotation | +5° (c=2 in methanol) |
Sensitivity | Hygroscopic |
Chemical Name or Material | L-Serine methyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'