missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ L Shaped Spreaders
Features smooth edges which avoids tearing and digging into agar
£35.65 - £494.00
Specifications
Color | Blue |
---|---|
Material | High-impact Polystyrene |
Sterility | Aseptically Manufactured |
Length (English) | 5.5 in. |
Length (Metric) | 140 mm |
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Product Type | Price | Quantity & Availability | ||||
15742125
|
Microspec™
E10 |
500 | Inoculation Product |
£35.65
Pack of 500 |
In Stock
Log in to see stock. |
||||
15752125
|
Microspec™
E4 |
500 | Spreader |
£494.00
Pack of 500 |
Estimated Shipment: 24-07-2024
Log in to see stock. |
||||
15732125
|
Microspec™
E1 |
7500 | Spreader |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- High Impact Polystyrene construction
- Full length to avoid contamination
- Disposable
- Sterile, aseptically manufactured
- Ideal for bacterial counts
Specifications
Blue | |
Aseptically Manufactured | |
140 mm |
High-impact Polystyrene | |
5.5 in. | |
Yes |